1-Propanesulphonyl chloride ≥97%

Leverandør: Thermo Scientific
Danger

Synonymer: Propane-1-sulphonyl chloride

H60747.AC H60747.AE
H60747.ACEA 730 NOK
H60747.AC H60747.AE
1-Propanesulphonyl chloride ≥97%
1-Propanesulphonyl chloride
Formel: C₃H₇ClO₂S
MW: 142,61 g/mol
Kokepunkt: 78…79 °C
Smeltepunkt: –46 °C
Tetthet: 1,267
Flash punkt: 97 °C (207 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007462
CAS-nummer: 10147-36-1
EINECS: 233-414-7
FN: 3265
ADR: 8,II

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR