2-Chloroethyl vinyl ether ≥97.0% (by GC) stabilised
Leverandør: TCI
|
Danger
|
Stabilisering:
Stabilised with 4-methoxyphenol (MEHQ)
C0174-25ML
C0174-500ML
TCIAC0174-25MLEA
1225
NOK
TCIAC0174-25ML
TCIAC0174-500ML
2-Chloroethyl vinyl ether ≥97.0% (by GC) stabilised
2-Chloroethyl vinyl ether
Formel:
H₂CCHOCH₂CH₂Cl MW: 106,55 g/mol Kokepunkt: 108°C Smeltepunkt: –70°C Tetthet: 1,0525 g/cm³ (15 °C) Flash punkt: 27°C |
MDL nummer:
MFCD00000973 CAS-nummer: 110-75-8 EINECS: 203-799-6 FN: 1992 ADR: 3,III Merck Indeks: 14,02139 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...