3-Hydroxy-5-methylisoxazole ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
H1348-5G
H1348-25G
TCIAH1348-5GEA
460
NOK
TCIAH1348-5G
TCIAH1348-25G
3-Hydroxy-5-methylisoxazole ≥98.0% (by GC)
3-Hydroxy-5-methylisoxazole
Formel:
C₄H₅NO₂ Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD00144468 CAS-nummer: 10004-44-1 EINECS: 233-000-6 Merck Indeks: 14,04856 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 84.0 to 88.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...