4-Fluoro-o-anisaldehyde ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
4-Fluoro-2-methoxybenzaldehyde, 4-Fluoro-2-anisaldehyde
F0827-5G
F0827-1G
TCIAF0827-5GEA
1925
NOK
TCIAF0827-5G
TCIAF0827-1G
4-Fluoro-o-anisaldehyde ≥97.0% (by GC)
4-Fluoro-o-anisaldehyde
Formel:
C₈H₇FO₂ MW: 154,14 g/mol |
MDL nummer:
MFCD00143318 CAS-nummer: 450-83-9 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 97.0 % |
Melting point | 59.0 to 62.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...