4-(4-Hydroxyphenyl)cyclohexanone ≥95.0% (by GC)

Leverandør: TCI

H1316-5G H1316-25G
TCIAH1316-5GEA 395 NOK
TCIAH1316-5G TCIAH1316-25G
4-(4-Hydroxyphenyl)cyclohexanone ≥95.0% (by GC)
4-(4-Hydroxyphenyl)cyclohexanone
Formel: C₁₂H₁₄O₂
MW: 190,24 g/mol
MDL nummer: MFCD00210693
CAS-nummer: 105640-07-1
EINECS: 423-920-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8463f704dc0b4ebae1c2d86d060dc8e06c78c7bb&vid=8463f704dc0b4ebae1c2d86d060dc8e06c78c7bb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 168.0 to 172.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR