Monomethyl succinate 95%

Leverandør: Thermo Scientific

Synonymer: 4-Methoxy-4-oxobutanoic acid, Methyl hydrogen succinate, Succinic acid monomethyl ester

H59722.14 H59722.22
H59722.14EA 1075 NOK
H59722.14 H59722.22
Monomethyl succinate 95%
Monomethyl succinate
Formel: C₅H₈O₄
MW: 132,11 g/mol
Kokepunkt: 151 °C (20 mmHg)
Smeltepunkt: 54…57 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002788
CAS-nummer: 3878-55-5
EINECS: 223-408-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR