Microtube racks, isofreeze, reversible, with lid
Leverandør: DELTALAB
M-570 M-571
479-4270EA 6350 NOK
479-4270 479-4275
Microtube racks, isofreeze, reversible, with lid
Stativer for rør Kjølestativ
PC.
  • Filled with a non-toxic gel
  • Choice of 0 or –10 °C versions
  • Transparent lid