Millex® Syringe Filter Units, Hydrophilic PTFE Membrane
Leverandør: Merck
SLCR025NS SLLGC13NL SLLHR04NL SLLGR04NL SLCR025NK SLLGC25NS SLLHC25NS SLLGH25NK SLLGH25NS SLLHC13NL SLLGH25NB
514-0407EA 0 NOK
514-0407 514-0448 514-0451 514-0449 MILFSLCR025NK MILFSLLGC25NS MILFSLLHC25NS MILFSLLGH25NK MILFSLLGH25NS MILFSLLHC13NL MILFSLLGH25NB
Millex® Syringe Filter Units, Hydrophilic PTFE Membrane
Filter Sprøytefilter
HPLC certified filters for the filtration of organic and aqueous solutions in HPLC.
  • Low protein binding and high chemical resistance
Tested and certified for UV-absorbing substances at wavelengths of 210 and 254 nm with water and acetonitrile; ensuring consistent quality between batches.
Inlet/outlet: Luer lock female/Luer male.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Fyllingsmengde 100 ml (<100 μl)
Maks. bemanningstrykk 45 °C