Fluid aspiration systems, WOB-L®
Leverandør: Welch by Gardner Denver
U2515C-75 U2511C-75
ILMVU2515C-75EA 19775 NOK
ILMVU2515C-75 ILMVU2511C-75
Fluid aspiration systems, WOB-L®
Pumper Fatpumper
These heavy-duty diaphragm pumps are specially designed for use in suction and filtration.
The connections with the pump are made by means of a quick plug.
Leveringsinformasjon: System includes an autoclavable vessel, adapters, tube, hydrophobic filters, control vacuum and manometer.
Bestill nå
SPESIFIKASJONER
Modell WOB-L® 2515 WOB-L® 2511
Environmentally Preferable
Gjennomstrømningshastighet 28000 (at 50 Hz)
34000 (at 60 Hz)
9200 (at 50 Hz)
11000 (at 60 Hz)
Støynivå 46 dB(A) 45 dB(A)