Sample container with screw cap
Leverandør: DELTALAB
409511 409711 409702 409726 409701 409726.G
216-4051EA 3400 NOK
216-4051 216-4052 216-4100 DELT409726 DELT409701 DELT409726.G
Sample container with screw cap
Prøvecontainere Beholdere, generelt bruk
Container PP, natural or blue, with PE screw cap.
  • With or without label
  • Blue containers are ideal for photosensitive samples
  • Bulk or individually wrapped
To store and transport samples securely.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904