Sample containers, with screw cap
Leverandør: DELTALAB
409752 409756 409756.B
DELT409752EA 3925 NOK
DELT409752 DELT409756 DELT409756.B
Sample containers, with screw cap
Prøvecontainere Beholdere, generelt bruk
Ultra clear PP containers with PE leak proof screw cap and PS spoon.
  • Graduated in 100 ml increments
  • Frosted panel on side for easy writing
  • Good chemical and temperature resistance
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904