Calcein-AM cell viability assay kits
Leverandør: Biotium
30002 30002-T 30026
30002.EA 5400 NOK
30002. 30002-T 30026.
Calcein-AM cell viability assay kits
Assay Cellulær analyse
Calcein-AM is a non-fluorescent, membrane permeable compound. Esterase activity in the cytoplasm of viable cells converts calcein-AM to the green fluorescent, membrane impermeant compound calcein, which is retained in viable cells with intact plasma membranes. The Viability/Cytotoxicity Assay kit for Animal Live & Dead Cells pairs calcein-AM with the vital dye ethidium homodimer III for quantitation of live and dead cells.