Volumetric flasks, class A, amber, white markings, DURAN®
Leverandør: DWK Life Sciences
246760954 246761059 246761256 246761453 246761753 246762458 246762552 246763257 246763651 246764459 246765455 246766357
612-5293EA 1125 NOK
612-5293 612-5294 612-5295 612-5296 612-5297 612-5298 612-5299 612-5300 612-5309 612-5316 612-5321 612-5322
Volumetric flasks, class A, amber, white markings, DURAN®
Kolber Målekolber
DURAN®, borosilicate glass 3.3, amber, with PE stopper, conformity certified.
  • White markings, with one graduation mark and new ergonomic polyethylene stopper
  • Certificate of conformity 'DE-M'
  • With batch certificate
*Wide neck.
With printed retrace code (batch identification).
Sertifiseringer: ISO 1042