Disinfectants, ClearKlens Cleansinald, sterile
Leverandør: Diversey
100848254 100848253 7513417 7516429
DIVY100848254EA 4150 NOK
DIVY100848254 DIVY100848253 DIVY7513417 141-0893
Disinfectants, ClearKlens Cleansinald, sterile
Rengjøringsløsninger Desinfeksjon
Surface active disinfectant based on a blend of quaternary ammonium and alkyl amines.
  • 0,2 µm filtered, aseptically filled and double bagged
  • Broad spectrum of activity
  • Supplied with certificate of analysis and sterility where applicable
  • Compatible with ClearKlens Tego® 2000, ClearKlens Tego® 2001 and ClearKlens Bi-Spore as a rotational process
Sertifiseringer: Passes CEN 1276, CEN 1650 and CEN 13697
NB: Use biocides safely. Always read the label and product information before use.
Articles may not be available in all countries. Please contact your local Avantor office.