Slip-resistant fine resilient tapes and treads, 200 series, Safety-Walk™
Leverandør: 3M
SW3WS25 SW3TR50 SW3WS50 3WS100NN SW3TR122N SW3TR25N
MMMASW3WS25EA 5825 NOK
MMMASW3WS25 MMMASW3TR50 MMMASW3WS50 MMMA3WS100NN MMMASW3TR122N MMMASW3TR25N
Slip-resistant fine resilient tapes and treads, 200 series, Safety-Walk™
Tape
High friction, fine resilient textured, slip-resistant material.
  • For wet, barefoot-traffic areas
  • Non-mineral, fine-textured, slip-resistant vinyl surface
  • High performance, water-resistant, acrylate adhesive is ideal for wet conditions
  • Custom shapes and sizes are available
Fine-textured, slip-resistant surface with a pressure-sensitive adhesive backing provides enhanced safety. Reverse side coated with pressure sensitive adhesive protected by a removable liner. Designed for use in wet, barefoot traffic areas. Applications include use on showers, tubs,More Product Information