Volumetric flasks, class B, white markings, DURAN®
Leverandør: DWK Life Sciences
246703251 246702555 246700957 246701053 246701259 246701456 246707356 246706351 246705458 246704453 246703654 246701756
612-5277EA 830 NOK
612-5277 612-5276 612-5271 612-5272 612-5273 612-5274 612-5282 612-5281 612-5280 612-5279 612-5278 612-5275
Volumetric flasks, class B, white markings, DURAN®
Kolber Målekolber
DURAN®, borosilicate glass 3.3, clear, with PE stopper.
  • With one graduation mark
  • Ergonomic stopper
  • Good chemical resistance, good temperature resistance
Error limits according to class B, DIN EN ISO 1042.
*Vid hals.