Dispensere, Enkanals mikro
Leverandør: DRUMMOND
3-000-310 3-000-320 3-000-325 3-000-350 3-000-375 3-000-385 3-000-388 3-000-390
DRUM3-000-310EA 11725 NOK
DRUM3-000-310 DRUM3-000-320 DRUM3-000-325 DRUM3-000-350 DRUM3-000-375 DRUM3-000-385 DRUM3-000-388 DRUM3-000-390
Dispensere, Enkanals mikro
Pipetter Micropipettes
Highly accurate micro-dispensers with borosilicate glass bores and stainless steel plunger.
  • Overall volumetric accuracy of ±1% (at full volume)
  • PTFE tips on all plungers 25 µl or larger
  • Precision molded plastic bores on 1000 μl model