CO₂, humidity and temperature data loggers
Leverandør: ROTRONIC INSTRUMENTS
CL11 CP11
620-2320EA 4950 NOK
620-2320 620-2321
CO₂, humidity and temperature data loggers
Dataloggere
These multiple parameter meters simultaneously measure and record CO₂, humidity and temperature. Use the SW2.1 software to program, download data and save logged files.
  • Maximum, minimum and average value display
  • Display with backlight
  • Mini USB port for connection to a PC
  • HYGROMER® IN-1 humidity sensor
  • Calculate dew point and wet bulb temperature (only CP11)
Adjustable audible and visual CO₂ alarm 40000 data point memory for CO₂, humidity and temperature values.
Leveringsinformasjon: The CL11 is delivered with an AC adapter. Both devices areMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Grensesnitt USB USB
levetid batteri 400 days 400 days
Vekt 200 g 290 g
B×D×H 120×45×157 mm 234×77×42 mm
Dataminne - Antall avlesninger 40000 18000