Examination gloves, vinyl, Peha-soft® Vinyl Powderfree
Leverandør: HARTMANN PAUL
9421708 9421718
HART9421708EA 125 NOK
HART9421708 HART9421718
Examination gloves, vinyl, Peha-soft® Vinyl Powderfree
Hansker
These hard-wearing disposable gloves can be used in all areas where the staff is vulnerable to contamination. Especially suitable for people who are hypersensitive to glove powder or with allergies to latex proteins (Type I allergy) or accelerators (type IV allergy).
  • Non sterile, powder-free, smooth finish
  • Latex-free
  • With an inner polymer lining for easy donning
  • Resilient, tear resistant and safe
  • Good fit and excellent tactile sensitivity
Sertifiseringer: EN 455; AQL 1,5
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Ambidekstral/håndspesifikk Ambidextrous
Engangs/flergangs engangs
Latex/latexfri Latex-fri
Pudder/pudderfri uten pudder
Overflate Smooth
Tykkelse mansjett 0,07 mm
Tykkelse finger 0,15 mm
Mansjett tykkelse 0,10 mm
EN norm EN 455