HPLC columns, MAbPac™ HIC-20
Leverandør: Thermo Fisher Scientific
088553 088554 088555
555-7357EA 16325 NOK
555-7357 555-7358 555-7359
HPLC columns, MAbPac™ HIC-20
Kromatografikolonner
The MAbPac™ HIC-20 HPLC column are designed for separation of monoclonal antibodies (mAbs), mAb fragments and oxidised variants by hydrophobic interaction chromatography (HIC).
  • High resolution and high efficiency
  • Compatible with both organic solvent and aqueous mobile phase
  • Rugged column stability
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
pH område 2 - 9
Temperaturgrense 60 °C