VWR®, Sampling Scoops
Leverandør: VWR Collection
231-0490EA 3050 NOK
231-0490 231-0491 231-0622 231-0623 231-0624 231-0625
VWR®, Sampling Scoops
Øser
PS, white. These sterile scoops are ideal for sampling powder, granules and pastes.
  • Individually packed
  • Sterilised by gamma radiation
Thanks to the special handle construction, and the slightly angled edge, the scoop stands horizontally and can be laid down without the material trickling out.
Sertifiseringer: Production, assembly and packaging according to cleanroom Class 7 (10000). According to EU food and FDA guidelines.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904.