Hibar® HPLC Columns, with Purospher® Sorbents
Leverandør: Merck
1.00423.7130 1.00424.7130 1.50009.7130 1.50012.7130 1.50013.7127 1.50013.7130 1.50013.7184 1.50182.7127 1.50182.7130 1.50183.7127 1.50183.7130 1.50009.7184 1.50183.7185 1.50182.7174 1.00423.7184 1.50009.7220 1.00424.7184 1.50183.7177 1.00424.7175 1.00424.7194 1.50182.7185 1.50009.7174 1.00424.7177 1.50013.7180 1.50013.7175 1.00423.7185 1.50183.7131 1.50183.7174 1.50183.7180 1.50183.7194 1.51196.7131 1.50182.7177 1.51196.7174 1.51196.7175 1.51196.7177 1.51196.7180 1.51196.7194 1.51196.7220 1.00424.7174 1.00424.7180 1.50182.7194 1.50012.7185 1.50012.7194 1.50013.7194 1.50012.7175 1.50012.7177 1.50012.7180 1.50012.7220 1.50013.7174 1.50013.7177 1.50013.7220 1.50182.7184 1.00424.7185 1.50182.7175 1.50182.7180 1.50182.7220 1.50012.7174 1.50012.7184
1.00423.7130EA 12200 NOK
1.00423.7130 1.00424.7130 1.50009.7130 1.50012.7130 1.50013.7127 1.50013.7130 1.50013.7184 1.50182.7127 1.50182.7130 1.50183.7127 1.50183.7130 1.50009.7184 1.50183.7185 1.50182.7174 1.00423.7184 1.50009.7220 1.00424.7184 1.50183.7177 1.00424.7175 1.00424.7194 1.50182.7185 1.50009.7174 1.00424.7177 1.50013.7180 1.50013.7175 1.00423.7185 1.50183.7131 1.50183.7174 1.50183.7180 1.50183.7194 1.51196.7131 1.50182.7177 1.51196.7174 1.51196.7175 1.51196.7177 1.51196.7180 1.51196.7194 1.51196.7220 1.00424.7174 1.00424.7180 1.50182.7194 1.50012.7185 1.50012.7194 1.50013.7194 1.50012.7175 1.50012.7177 1.50012.7180 1.50012.7220 1.50013.7174 1.50013.7177 1.50013.7220 1.50182.7184 1.00424.7185 1.50182.7175 1.50182.7180 1.50182.7220 1.50012.7174 1.50012.7184
Hibar® HPLC Columns, with Purospher® Sorbents
Kromatografikolonner
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of Purospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number MERC1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.