Anti-GTF2H1 Rabbit Polyclonal Antibody

Leverandør: Biorbyt
ORB1824-100 ORB1824-200
BIRBORB1824-100EA 0 NOK
BIRBORB1824-100 BIRBORB1824-200
Anti-GTF2H1 Rabbit Polyclonal Antibody
Antistoffer
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: GTF2H1 (general transcription factor IIH, polypeptide 1, 62kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR