Anti-BEST1 Rabbit polyclonal antibody

Leverandør: Biorbyt
ORB2889-100 ORB2889-200
BIRBORB2889-100EA 0 NOK
BIRBORB2889-100 BIRBORB2889-200
Anti-BEST1 Rabbit polyclonal antibody
Antistoffer
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: BEST1 (bestrophin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR