Anti-IL10RA Rabbit Polyclonal Antibody
Leverandør: Biorbyt
ORB13490-100 ORB13490-200
BIRBORB13490-100EA 0 NOK
BIRBORB13490-100 BIRBORB13490-200
Anti-IL10RA Rabbit Polyclonal Antibody
Antistoffer
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: IL10RA (interleukin 10 receptor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå