Anti-IL12RB1 Rabbit Polyclonal Antibody
Leverandør: Biorbyt
ORB13492-100 ORB13492-200
BIRBORB13492-100EA 0 NOK
BIRBORB13492-100 BIRBORB13492-200
Anti-IL12RB1 Rabbit Polyclonal Antibody
Antistoffer
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: IL12RB1 (interleukin 12 receptor, beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat