Anti-NLRP4 Rabbit Polyclonal Antibody

Leverandør: Biorbyt
ORB100982-100 ORB100982-200
BIRBORB100982-100EA 0 NOK
BIRBORB100982-100 BIRBORB100982-200
Anti-NLRP4 Rabbit Polyclonal Antibody
Antistoffer
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: NLRP4 (NLR family, pyrin domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR