Anti-IgG Rabbit Polyclonal Antibody (CF™ 647)

Leverandør: Biotium
20049 20049-1
20049.EA 3700 NOK
20049. 20049-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 647)
Antistoffer
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 647 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Goat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR