Anti-IgG Goat Polyclonal Antibody (CF™ 640R)

Leverandør: Biotium
20085-1 20085
20085-1EA 1500 NOK
20085-1 20085.
Anti-IgG Goat Polyclonal Antibody (CF™ 640R)
Antistoffer
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Guinea pig
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR