Anti-SCARB1 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
NB400-104 NB400-104SS
NOVUNB400-104EA 8600 NOK
NOVUNB400-104 NOVUNB400-104SS
Anti-SCARB1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SCARB1 (scavenger receptor class B, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR