Anti-PDPN Golden Syrian Hamster Monoclonal Antibody [clone: 8.1.1]

Leverandør: Novus Biologicals
NB600-1015 NB600-1015SS
NOVUNB600-1015EA 7675 NOK
NOVUNB600-1015 NOVUNB600-1015SS
Anti-PDPN Golden Syrian Hamster Monoclonal Antibody [clone: 8.1.1]
Antistoffer
Type: Primary
Antigen: PDPN (podoplanin)
Clonality: monoclonal
Clone: 8.1.1
Conjugation: unconjugated
Epitope:
Host: Golden Syrian Hamster
Isotype: IgG
Reactivity: Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol PDPN
Antigen-navn Podoplanin
Antigen synonymer T1A-2|36-KD|glycoprotein|PA2.26 antigen|hT1alpha-2|hT1alpha-1|AGGRUS|T1A|HT1A-1|GP36|Gp38|PA2.26|CTA-520D8.1|lung type-I cell membrane-associated glycoprotein (T1A-2)|T1-alpha|lung type I cell membrane associated glycoprotein|glycoprotein 36|OTS8|GP40
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon 8.1.1
Reaktivitet Mouse
Vert Golden syrian hamster
Gen-ID 10630
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Immunutfelling Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR