Anti-KDM5B Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
NBP1-97310 NBP1-97310SS
NOVUNBP1-97310EA 7125 NOK
NOVUNBP1-97310 NOVUNBP1-97310SS
Anti-KDM5B Rabbit Polyclonal Antibody
Antistoffer
Rabbit Polyclonal JARID1B Antibody. Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (-). Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: JARID1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol JARID1B
Antigen-navn lysine (K)-specific demethylase 5B
Environmentally Preferable
Antigen synonymer PLU1|JARID1B|CT31|RBP2-H1|PUT1|RBBP2H1A|PPP1R98|PLU-1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Gen-ID 10765
Isotype
Western blot Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR