Anti-CB018 Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
USBI033230-BIOTINEA 10475 NOK
USBI033230-BIOTIN
Anti-CB018 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: CB018
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR