Anti-HIGD2A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: US Biological
USBI036556-APEA 10475 NOK
USBI036556-AP
Anti-HIGD2A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: HIGD2A (HIG1 hypoxia inducible domain family, member 2A)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR