Anti-LGALSL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: US Biological
USBI036811-HRPEA 10475 NOK
USBI036811-HRP
Anti-LGALSL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: LGALSL (lectin, galactoside-binding-like)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR