Anti-SQSTM1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverandør: United States Biological
USBI042284-HRPEA 10475 NOK
USBI042284-HRP
Anti-SQSTM1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Dot blot 1:500
Type: Primary
Antigen: SQSTM1 (sequestosome 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human