Anti-CERS2 Mouse Polyclonal Antibody

Leverandør: United States Biological
USBI129010EA 6425 NOK
USBI129010
Anti-CERS2 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CERS2 (ceramide synthase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR