Anti-BID Goat polyclonal antibody unconjugated

Leverandør: United States Biological
USBIB1095-03FEA 6775 NOK
USBIB1095-03F
Anti-BID Goat polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: BID (BH3 interacting domain death agonist)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR