Anti-CDH17 Mouse Monoclonal Antibody [clone: 3H2]

Leverandør: United States Biological
USBIC0107-17AEA 8650 NOK
USBIC0107-17A
Anti-CDH17 Mouse Monoclonal Antibody [clone: 3H2]
Antistoffer
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CDH17 (cadherin 17, LI cadherin (liver-intestine))
Clonality: Monoclonal
Clone: 3H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR