Anti-LGALS3 Rabbit Polyclonal Antibody

Leverandør: United States Biological
USBIG1044-50MEA 8300 NOK
USBIG1044-50M
Anti-LGALS3 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:5000; Western Blot: 1:500-1:1000; Immunofluorescence: 1:100-1:500; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: LGALS3 (lectin, galactoside-binding, soluble, 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR