Anti-KPNA1 Mouse Monoclonal Antibody [clone: 2A4-1B5]

Leverandør: United States Biological
USBIK0102-02GEA 8650 NOK
USBIK0102-02G
Anti-KPNA1 Mouse Monoclonal Antibody [clone: 2A4-1B5]
Antistoffer
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: KPNA1 (karyopherin alpha 1 (importin alpha 5))
Clonality: monoclonal
Clone: 2A4-1B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR