Anti-MAP Kinase 1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverandør: United States Biological
USBIM2351-80BEA 6475 NOK
USBIM2351-80B
Anti-MAP Kinase 1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA:0.25 ?g/ml; Western Blot:1 ?g/ml
Type: Primary
Antigen: MAP Kinase 1
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human