Anti-Nectin 1 Mouse Monoclonal Antibody [clone: 6A139]

Leverandør: United States Biological
USBIN2167EA 8550 NOK
USBIN2167
Anti-Nectin 1 Mouse Monoclonal Antibody [clone: 6A139]
Antistoffer
Recommended dilution: Immunoprecipitation: 5-10 ?g/reaction; Western Blot: 0.1-1 ?g/ml; Flow Cytometry: 5-10 ?g/ml; ELISA: 0.1-1 ?g/ml

Type: Primary
Antigen: Nectin 1
Clonality: monoclonal
Clone: 6A139
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR