Anti-Staphylococcus aureus Enterotoxin B Rabbit Polyclonal Antibody

Leverandør: United States Biological
USBIS7965-35QEA 19800 NOK
USBIS7965-35Q
Anti-Staphylococcus aureus Enterotoxin B Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Staphylococcus aureus Enterotoxin B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR