Biological safety cabinet, class II, type A2, Purifier® Logic®+
Leverandør: LABCONCO
302481030 302481150 302611040 302681150 302411030 302581041
LBCP302481030EA 0 NOK
LBCP302481030 LBCP302481150 LBCP302611040 LBCP302681150 LBCP302411030 LBCP302581041
Biological safety cabinet, class II, type A2, Purifier® Logic®+
Avtrekksskap og kabinetter Sikkerhetskabinetter
Purifier Logic+ Class II, Type A2 Biosafety Cabinets provide personnel, product and environmental protection from hazardous particulates such as agents that require Biosafety Level 1, 2 or 3 containment. Other appropriate applications include work with antineoplastic drugs, genetic material, asbestos and additional substances that generate hazardous airborne particulates. These cabinets are designed to work with antineoplastic drugs, genetic material, asbestos and additional substances that generate hazardous airborne particulates. When canopy connected, the biosafety cabinets may be usedMore Product Information
Bestill nå
SPESIFIKASJONER
Modell Purifier® Logic®+ 4 feet, with base stand Purifier® Logic®+ 6 feet, without base stand Purifier® Logic®+ 6 feet, with base stand Purifier® Logic®+ 4 feet, without base stand Purifier® Logic®+ 5 feet, with base stand
Environmentally Preferable Energy Efficient,Sustainable Materials Energy Efficient,Sustainable Materials Energy Efficient,Sustainable Materials Energy Efficient,Sustainable Materials Energy Efficient,Sustainable Materials
Elektrisk tilkobling 230 V, 50/60 Hz, 6 A 230 V, 50/60 Hz, 8 A 230 V, 50/60 Hz, 8 A 230 V, 50/60 Hz, 6 A 230 V, 50/60 Hz, 8 A
Ext. W×D×H 1379×792×2266 mm 1989×792×1567 mm 1989×792×2266 mm 1379×792×1567 mm 1684×792×1567 mm
Weight 304 kg 365 kg 410,5 kg 265 kg 324 kg