Shodex HILICpak VG-50 HILIC Columns for Polar Compounds and Sugar Analysis
Leverandør: Resonac
F6711100 F7630100 F7630200 F6711200 F7630300
554-4574EA 7600 NOK
554-4574 554-4572 554-4573 SHOWF6711200 SHOWF7630300
Shodex HILICpak VG-50 HILIC Columns for Polar Compounds and Sugar Analysis
Kromatografikolonner
Robust polymer-based HILIC column for polar analytes and saccharides.
  • Polymer-based HILIC columns with tertiary amino (-NR2) groups
  • Suitable for polar analytes and saccharide analysis using HILIC mode
  • Recovers reducing saccharides with high ratio (improved results for galactose, mannose, arabinose and xylose in comparison to Asahipak NH2P-50 series)
  • Polymer-based packing material provides excellent chemical stability and minimum deterioration over an extended time period
  • pH stability: 2 to 13 (silanol free)
  • Easily regenerated by washing in an alkaline
More Product Information
Bestill nå
SPESIFIKASJONER
Beskrivelse HILICpak VG-50G 4A HILICpak VG-50 4E HILICpak VG-50 4D HILICpak VG-50G 2A HILICpak VG-50 2D
Environmentally Preferable
Maks. trykk 150 bar 15 MPa (150 bar) 10 MPa (100 bar) 15 MPa (150 bar)
pH område 2 - 13 2 - 13 2 - 13 2 - 13 2 - 13
Temperaturgrense 4 - 60 °C 4 - 60 °C 4 - 60 °C 4 - 60 °C 4 - 60 °C