Hansker, Neopren, Microflex® 73-847
Leverandør: Ansell
73-847/5.5-6 73-847/7.5-8 73-847/8.5-9 73-847/6.5-7 73-847/9.5-10
112-4371EA 625 NOK
112-4371 112-4373 112-4374 112-4372 112-4375
Hansker, Neopren, Microflex® 73-847
Hansker
These disposable neoprene gloves provide chemical and liquid protection.
  • Soft and pliable neoprene formulation provides a more comfortable wear for longer periods, while allowing workers greater tactile sensitivity
  • Textured fingers ensure a strong and secure grip, while the advanced formulation helps deliver excellent wet and dry grip
  • Proven to reduce muscle effort and hand fatigue that can lead to serious injuries while enhancing gripping abilities in wet conditions
  • Made without natural rubber proteins and powder to help protect workers from allergies,
More Product Information
Bestill nå
SPESIFIKASJONER
Modell 73-847
Environmentally Preferable
Ambidekstral/håndspesifikk Ambidextrous
Engangs/flergangs engangs
Latex/latexfri Latex-fri
Pudder/pudderfri uten pudder
Overflate Textured fingertips
Type mansjett Perlebesatt
EN norm EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS