Filter discs, biplane, series 16, porosity 1, VitraPOR®
Leverandør: ROBU
VitraPOR®
ROBU16121EA 2750 NOK
ROBU16121 ROBU16301 ROBU62001 ROBU16201 ROBU16101 ROBU62101 ROBU62201 ROBU62301 ROBU62401 ROBU62501 ROBU62601 ROBU62701 ROBU62801 ROBU62901 ROBU61901 ROBU61801 ROBU61701 ROBU61601 ROBU61501 ROBU61401 ROBU61301 ROBU16251 ROBU63001 ROBU16111 ROBU63501 ROBU63801 ROBU16701 ROBU16601 ROBU16501 ROBU16401 ROBU16901 ROBU16801
Filter discs, biplane, series 16, porosity 1, VitraPOR®
Filter Filterdisker
The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
  • Very good chemical resistance
  • Very good temperature resistance
Made of borosilicate glass. The faces of filter discs from the series 16 are precisely ground down to tight tolerances and plane surfaces as may be required in specific installation circumstances.