Quartz microfibre filters for air sampling, grade QM-A, Whatman™
Leverandør: Whatman products (Cytiva)
1851-025 1851-037 1851-047 1851-050 1851-055 1851-085 1851-865 1851-150 1851-032 1851-082 1851-090 1851-101 1851-110 1851-118 1851-8866 1851-045 1851-037_L
512-0430EA 5200 NOK
512-0430 516-2608 516-2611 516-5116 516-0199 516-5119 512-0431 516-5122 513-0112 512-0671 513-0086 513-0113 516-5120 516-5121 516-5123 WHAT1851-045 WHAT1851-037_L
Quartz microfibre filters for air sampling, grade QM-A, Whatman™
Filter Glassfiberfilter
These high purity quartz microfibre filters are used in many air sampling filter applications, including sampling of acidic gases, stacks, flues and aerosols. They are suitable for PM-10 sampling and analysis.
  • Binder-free
  • High loading capacity
Because of the low level of alkaline earth metals, 'artifact' products of sulfates and nitrates (from SO₂ and NO₂) are virtually eliminated. High-purity quartz microfiber filters are therefore applicable for particulate monitoring and heavy metal analysis.
Bestill nå
SPESIFIKASJONER
Nr. QM-A QM-A
Environmentally Preferable
Vekt 85 g/m² 85 g/m² 85 g/m² 85 g/m²
Thickness 475 µm 475 µm 475 µm 475 µm
Partikkelretensjon 2,2 µm 2,2 µm 2,2 µm 2,2 µm
Luftgjennomstrømning 6,3 s/100 ml/6,5 cm² 6,3 s/100 ml/6,5 cm²
Maks. temp omgivelser 800 °C 800 °C 800 °C 800 °C