Avantor® ACE® C4-300, HPLC Columns, 3 µm
Leverandør: Avantor
ACE-213-0302EA 0 NOK
ACE-213-0302 ACE-213-0502 ACE-213-0503 ACE-213-0546 ACE-213-1001 ACE-213-1002 ACE-213-1003 ACE-213-1046 ACE-213-1502 ACE-213-1503 ACE-213-1546 ACE-213-0102GD ACE-213-0103GD ACE-213-10005 ACE-213-15001 ACE2131046MVK ACE2131546MVK ACETACE-213-2546
Avantor® ACE® C4-300, HPLC Columns, 3 µm
Kromatografikolonner
Avantor® ACE® C4-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
  • Designed for peptide and protein separations
  • Method validation kits contain three different batches of the same phase - available on request
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Karbonbelastning 2,6% 2,6%
Kappet ved endene Ja Ja Ja
Maks. trykk 275 bar 275 bar 275 bar
pH område 2 - 8 2 - 8 2 - 8
Overflate areal 100 m²/g 100 m²/g 100 m²/g
USP kode L26 L26 L26
Miljøsertifikat ISO 14001:2015 (530840 UM15) ISO 14001:2015 (530840 UM15) ISO 14001:2015 (530840 UM15)